Biopsy processing/embedding cassettes, Swingsette®, M516
Leverantör: Simport Scientific
M516-10 M516-11 M516-12 M516-2 M516-3 M516-4 M516-5 M516-6 M516-7 M516-8 M516-9
720-1642EA 3850 SEK
720-1642 720-1643 720-1644 720-1648 720-1649 720-1650 720-1651 720-1652 720-1653 720-1654 720-1655
Biopsy processing/embedding cassettes, Swingsette®, M516
Histologikassetter Biopsikassetter
Acetal polymer.
  • Integral hinged lid, with large tab for easier opening
  • Disposable plastic cassettes are highly resistant to histological solvents
  • Small, square openings ensure good fluid exchange without having to use foam pads or biopsy bags
  • Front face inclined 45° enabling use with certain automatic labelling systems
Förpackning: Each case contains three boxes of 500 cassettes.