Tissue cassettes in QuickLoad™ taped stacks, Unisette™, M505T
Leverantör: Simport Scientific
M505-10T M505-11T M505-12T M505-2T M505-3T M505-4T M505-5T M505-6T M505-7T M505-8T M505-9T
720-1389EA 2625 SEK
720-1389 720-1390 720-1391 720-1392 720-1393 720-1394 720-1395 720-1396 720-1397 720-1398 720-1399
Tissue cassettes in QuickLoad™ taped stacks, Unisette™, M505T
Histologikassetter Histokassetter
Moulded from a special high density acetal polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.
  • M506 series is similar to M505 but specially designed to hold biopsy specimens during the embedding process
  • Available in non cytotoxic, non metallic colours
  • The snap-latch and hinge-lock design prevents early separation of base and lid and allows for one handed operation
The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-pieceMore Product Information