Culture media preparator, MasterClave® 20
Leverantör: bioMérieux
4704390VC
710-1962EA 0 SEK
710-1962
Culture media preparator, MasterClave® 20
Mediaberedare
The Masterclave® 20 is an automated preparation system for culture media, agars or broths, and is designed for used by qualified, trained laboratory professionals in the clinical, pharmaceutical, food processing and cosmetics industries. With its high core temperature precision control, and high efficiency stirring system, the Masterclave® 20 system optimises work flow and guarantees homogeneity, fertility and sterility.
  • Capacity: 1 to 20 litres
  • Productivity: removes the preparatory stages such as medium stirring and bottle autoclaving
  • Reproducibility:
More Product Information