Test kits, photometric, Nitrogen total (Laton), LCK cuvette tests
Leverantör: Hach
LCK338 LCK238 LCK138
705-3014EA 2525 SEK
705-3014 705-3013 705-3012
Test kits, photometric, Nitrogen total (Laton), LCK cuvette tests
Testkit för miljöanalys
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
Certifikat: EN ISO 11905-1