Oxalsyra 0.1 M (0.2N) Eluent concentrate for IC, Supelco®

Leverantör: Merck

Synonyms: Ethanedioic acid

MQ100

68487-1L
68487-1LEA 928 SEK
68487-1L
Oxalsyra 0.1 M (0.2N) Eluent concentrate for IC, Supelco®
Oxalsyra
Formel: C₂H₂O₄
MW: 90.04 g/mol
Kokpunkt: 365
MDL Number: MFCD00002573
CAS nummer: 144-62-7
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c16338a23bec6416d4ae95b43b00156fa2979716&vid=755c7cdc9d7f83e2d06c217c6de7fe2f5f3c3fe5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR