3-metylpentan, Supelco®

Leverantör: Merck
Fara


MQ100

68320-50ML 68320-5ML
68320-50MLEA 4800 SEK
68320-50ML 68320-5ML
3-metylpentan, Supelco®
3-metylpentan
Formel: C₆H₁₄
MW: 86.18 g/mol
Kokpunkt: 64
Smältpunkt: –118 °C
Densitet: 0.664 g/cm³ (20 °C)
Flampunkt: –40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009342
CAS nummer: 96-14-0
EINECS: 202-481-4
UN: 1208
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=735c89dc450d547ebef567bb643698327134d388&vid=3022c34d07dad177b9a3300a8b67a94ff7182088&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR