Toluen ≥99.5%, AR®, Macron Fine Chemicals™

Leverantör: Avantor Performance Materials
Fara

Synonyms: Methylbenzene, Phenylmethane, FM approval

6784-45EA 6600 SEK
6784-45 6784-25
Toluen ≥99.5%, AR®, Macron Fine Chemicals™
Toluen
Formel: C₆H₅CH₃
MW: 92.14 g/mol
Kokpunkt: 110,6
Smältpunkt: –95 °C
Flampunkt: 4 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay (GC) ≥99.5%
Water (KF ≤0.03 %
Titrable Acid (meq/g) ≤0.0005
Residue after Evaporation ≤0.002 %
Sulfur Compounds ≤0.0003 %
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR