Supelco® TraceCERT® Organic Reference Standard, ʟ-Lysine monohydrochloride
Leverantör: Merck
67448-100MG
67448-100MGEA 887 SEK
67448-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Lysine monohydrochloride
Standarder Organic Standards
Certifikat: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43fe57832e4fbf300880526adfc953deab214a27&vid=43fe57832e4fbf300880526adfc953deab214a27&prid=web&clid=SAPDAM