Multiparameter Handheld Liquiline Mobile, CML18
Leverantör: Endress+Hauser
71550073 CML18-1035/0 CML18-1098/0 71631651
665-0564EA 15200 SEK
665-0564 665-0566 665-0567 ENDR71631651
Multiparameter Handheld Liquiline Mobile, CML18
Elektrokemimätare pH-mätare
Liquiline Mobile CML18 is a mobile multi-parameter handheld device for measuring various parameters (pH, oxygen, conductivity). Thanks to Memosens technology, sensors for the different parameters can be connected via inductive coupling (plug and play).
  • Easy operation: use your existing tablets and smartphones for operation and commissioning
  • Benefit from all the advantages of Memosens technology: Memosens sensors offer you the safest data transmission, the highest measured value availability and total ease of use
  • Trust your measured values: using the same
More Product Information