Conductivity standards, Potassium chloride, IUPAC
Leverantör: Hach
S51M001 S51M002 S51M003
665-0489EA 1100 SEK
665-0489 665-0490 665-0491
Conductivity standards, Potassium chloride, IUPAC
Standarder Konduktivitetsstandarder
Potassium chloride standard solutions according to OIML, traceable to NIST and Standard solution verified by Reference Materials Laboratory.
Each standard is delivered in an airtight package ensuring safe storage. The nominal value of the pH standard is guaranteed until first opening, for up to 2 years from date of manufacture.
Observed shelf life after opening, following recommended storage and use: 3 months.
Leveransinformation: Each standard is delivered in an airtight can and is supplied with a film-wrapped 'Certificate of Conformity and Traceability' as well as a DAkkS calibration certificate specifying the exact measured value and calculated expanded uncertainty (k=2) for each batch of conductivity solution.