Multi-parameter depth probes, MPP 930 IDS
Leverantör: WTW
401205 401206
665-0357EA 73400 SEK
665-0357 665-0358
Multi-parameter depth probes, MPP 930 IDS
Elektroder Elektroder, andra
These digital depth probes, with built-in barometric pressure and temperature compensation, combine dissolved oxygen, pH or ORP and conductivity measurements with accurate depth readings. They can be used with either one, two or three IDS electrodes. The multi-parameter meter Multi 3430 or Multi 3630 IDS is required for measurements.
  • No twisting, no screwing: Only a click is needed for creating a pressure, tension resistant, data proof connection to the MPP IDS
  • Optional special cable reel with sliding contacts for simultaneously winding the cable and
More Product Information