Specialelektrod för pH, InLab®
Leverantör: Mettler - Toledo
59904591 51343061 51343062 51343150 51343153 51343157 51343174 51343176 51343151 30248112 30248120 30248826 30248828 30244733 30253098
662-2893EA 5075 SEK
662-2893 662-2835 662-2836 662-2847 662-2848 662-2850 662-2869 662-2870 662-1904 662-2033 662-2034 662-2035 662-2036 662-2164 662-2040
Specialelektrod för pH, InLab®
Elektroder pH-elektroder
A variety of pH electrodes for use in different applications.
  • ARGENTHAL™ reference system: AgCl granulate-filled cartridge supplies the silver ions for the chemical reaction on the conducting wire; silver ion trap stops the Ag+ ions from discharging into the electrolyte solution, thus preventing contamination of the junction when using sulfides and proteins
  • SteadyForce™: Reference system in which the electrolyte is pressurised (3 bar) to ensure discharge of electrolyte even in viscous samples
  • MultiPin™: pH/temperature
Beställningsinformation: SuppliedMore Product Information