VWR®, Universella pH-elektroder
Leverantör: VWR Collection
662-1398EA 3300 SEK
662-1398 662-1383 662-1384 662-1385 662-1386 662-1387 662-1388 662-1389 662-1390 662-1391 662-1392 662-1393 662-1395 662-1382 662-1397 662-1399 662-1401 662-1402 662-1403 662-1404 662-1405 662-1406 662-1407 662-1396
VWR®, Universella pH-elektroder
Elektroder pH-elektroder
Laboratorie- och fältelektroder för allmänna tillämpningar. Levereras med en fast kabel eller stickpropp, VWR®-elektroder kan anslutas till de flesta pH-mätarna på marknaden.
  • Comfortable handling
  • Reliable, rapid readings
  • Gel or liquid electrolyte, glass or epoxy body shaft
  • Available with different connections: Plug head or fixed cable (1 metre) with either DIN or BNC type plug
All VWR® electrodes are checked at two points during the manufacturing process and then lot stamped to ensure quality. They are supplied with a soaker storage bottle and cap toMore Product Information
Order Now
SPECIFIKATIONER
Environmentally Preferable