Alcian blue 8GX 1% i ättiksyra 3% för mikroskopi, Sigma-Aldrich®

Leverantör: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ100

66011-100ML-F 66011-500ML-F
66011-100ML-FEA 686 SEK
66011-100ML-F 66011-500ML-F
Alcian blue 8GX 1% i ättiksyra 3% för mikroskopi, Sigma-Aldrich®
Alcian blue

Alcianblue 8GX solution has been used as a staining dye for chondrocytic, osteoblastic and adipocytic differentiation. It has also been used for the staining of sections in immunohistochemistry.


Alcian blue is a large conjugated dye molecule, and comprises of a central copper-containing pthalocyanine ring linked to four isothiouronium groups via thioether bonds. The isothiouronium groups are responsible for the cationic nature of alcian blue owing to them being moderately strong bases. It was initially used in dyeing of textile fibres. Alcian blue 8GX is a dye recommended normally for histological technique.

Formel: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
Densitet: 0,996…1,016 g/cm³
Storage Temperature: Room Temperature
MDL Number: MFCD00010720
CAS nummer: 75881-23-1

Order Now

Specifikation testresultat

Appearance (Colour) Blue to Very Dark Blue Blue/Blue-Green to Very Deep Blue/Blue-Green
Appearance (Form) Liquid
Density (D20/4) 0.996 - 1.016
Refractive index at 20 °C 1.331 - 1.341
Wavelength (1)(UV) 605 - 625 nm
Extinction (1) (UV) (1%/1 cm)
Solvent (UV) Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR