Low volume polarimeter tubes
Leverantör: BELLINGHAM STANLEY
35-71 35-72 35-73 35-74 35-80 35-81
635-0751EA 10525 SEK
635-0751 635-0752 635-0753 635-0754 BELL35-80 BELL35-81
Low volume polarimeter tubes
Polarimetrar
Low volume, short length, stainless steel and glass loaded PTFE tubes with low strain cover glasses and rubber washer for use with ADP 400 Series polarimeters or similar makes working in the visible spectrum. Sample may be injected using a Luer taper syringe or for automated flow-through systems, connected via suitable flexible tubing.
  • Standard 30 mm collar size (ICUMSA)
  • Stainless steel versions are available with a water jacket facilitating temperature controlled measurement using a waterbath
  • PTFE versions for aggressive samples
The design of sampleMore Product Information