Staining jars according to Coplin, WHEATON®
Leverantör: DWK Life Sciences
900570
631-2509EA 4150 SEK
631-2509
Staining jars according to Coplin, WHEATON®
Burkar Färgkyvetter
Soda-lime glass.
  • Linerless, white PP screw cap to reduce solvent evaporation
  • Capacity: Approx. 60 ml
This unit is used for staining slides, or as a developing chamber for thin layer chromatography. It holds five single 76×26 mm slides vertically, or 10 slides back to back.