Handhållna mikroskop, Dino-Lite® EarScope
Leverantör: Dino-Lite / IDCP B.V.
MEDL3E MEDL4E MEDL4EP
630-2607EA 3800 SEK
630-2607 630-2609 630-2610
Handhållna mikroskop, Dino-Lite® EarScope
Mikroskop
With this handheld microscope users can easily inspect and photograph the ear canal, the ear drum and the outer ear. Ideal for use in general practice, in Audiology and for ear, nose and throat specialists.
  • Detachable cap for hygienical reasons
  • Handy high quality details with sharp colours
  • Compact and reliable instrument for ear observation
The image can be viewed on a computer screen to simplify diagnosis simplifies and also allows the patient to watch the images as well. The images, either still photo or video, can be stored digitally and easilyMore Product Information