VWR® Traceable®, Dishwasher Thermometer
Leverantör: VWR Collection
620-9100EA 640 SEK
620-9100
VWR® Traceable®, Dishwasher Thermometer
Termometrar Digitaltermometrar
This thermometer ensures proper cleaning by providing proof that the required temperature of 82 °C is met during dishwashing. Designed for applications in wet laboratories, wash-down areas and outdoors. Used in food, pharmaceutical, cleanrooms, and industrial laboratories. Waterproof metal with internal stainless steel sensor.
  • Maximum/minimum values during a dishwashing cycle
Certifikat: Traceability to standards provided by NIST, complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Leveransinformation: Supplied with clip for attaching toMore Product Information