VWR® Traceable®, Digital Pocket Thermometer
Leverantör: VWR Collection
620-1902EA 720 SEK
620-1902
VWR® Traceable®, Digital Pocket Thermometer
Termometrar Digitaltermometrar
This digital thermometer is suitable for the monitoring of temperatures in lab equipment, plants and field use.
  • Shockproof
  • Fast response
  • One-switch operations for routine applications
K-type probe with beaded sensor, 120 cm cable.
Leveransinformation: Supplied with traceable certificate of calibration, probe with cable, leather case and battery.