Dispenser tips, PD-Tips II
Leverantör: Brand
705708 705710 705714 705736 705744 705706 705700 705718 705720 705730 705748 705712 705716 705742 705704 705738 705702 705732 705734 705740 705746
613-6215EA 649.3 SEK
613-6215 613-6216 613-6218 613-6225 613-6229 613-6214 613-6211 613-6220 613-6221 613-6222 613-6231 613-6217 613-6219 613-6228 613-6213 613-6226 613-6212 613-6223 613-6224 613-6227 613-6230
Dispenser tips, PD-Tips II
Pipettspetsar
The PD-Tips II dispenser tips for efficient, step-by-step dispensing enhance the proven features of the classic BRAND PD-Tips. Thanks to patented size coding, HandyStep® touch and HandyStep® electronic automatically detect the tip volume. The tips are also compatible with all other instruments of the HandyStep® family and with repetitive pipettes from other manufacturers.
  • Volumes from 0,1 to 50 ml
  • Cleanroom quality
  • Available in BIO-CERT® quality (free of DNA, RNase, endotoxins and ATP)
  • Manufactured from high-quality raw materials (cylinder: PP / Piston: PE-
More Product Information