Pipette controller, accu-jet® S
Leverantör: Brand
26351 26352 26353 26350 26361 26362 26363 26360
612-6818EA 3426.04 SEK
612-6818 612-6819 612-6820 612-6817 612-6822 612-6823 612-6824 612-6821
Pipette controller, accu-jet® S
Pipetter Pipettfyllare
The accu-jet® S pipette controller brings versatility and endurance to your team. It makes lab work simple, easy, and efficient, whether you work with cell cultures, in quality control or in a pharma lab.
  • Precise control for accurate volume setting and delicate cell culture work
  • Integrated rest position
  • Service friendly and sustainable
  • Ergonomic handgrip and perfectly balanced design
  • Fills a 25 ml pipette at maximum motor speed in only 3 seconds
  • Battery with intelligent charging technology and two colour charge indicators
  • Four attractive colours