VWR® Advanced, Exact Volume Transfer Pipettes, Non Sterile
Leverantör: VWR Collection
612-5846EA 260 SEK
612-5846 612-5847 612-5848 612-5849 612-5850
VWR® Advanced, Exact Volume Transfer Pipettes, Non Sterile
Pipetter Pasteurpipetter
LDPE, disposable. These unbreakable all-in-one-pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
  • Uses from –20 to +50 °C
  • Available non sterile and sterile
  • Meet the requirements of BSE/TSE
Ideal for transferring and dispensing liquids safely in all types of laboratories.
Pipettes are manufactured under the ISO 13485 standard.