VWR® TA Professional, Analytical Balances with Touch Screen
Leverantör: VWR Collection
611-3677EA 26325 SEK
611-3677 611-3684 611-3686 611-3683 611-3679 611-3681 611-3674 611-3676 611-3828 611-3829 611-3830 611-3831
VWR® TA Professional, Analytical Balances with Touch Screen
Vågar
Laboratory analytical balances that have 5" colour touch screen display with advanced applications and large databases for professional weighing. The user has easy and intuitive access to balance functions, via interactive, icon-driven menus ensuring efficient and productive weighing. Balances can be used by up to 10 different users, each with their own customisable settings and password protection. Ideal for laboratories requiring GLP compliance. All models have a stainless steel top pan.
  • Internal and external calibration (optional factory calibration
More Product Information