Valinomycin, Selectophore™, Funtion tested ionophore, Supelco®

Leverantör: Merck
Fara


MQ100

60403-100MG
60403-100MGEA 4410 SEK
60403-100MG
Valinomycin, Selectophore™, Funtion tested ionophore, Supelco®
Valinomycin

Important research tool for membrane studies; As ionophor in membrane electrodes for the determination of K+-activities.

Formel: C₅₄H₉₀O₁₈N₆
MW: 1111.34 g/mol
MDL Number: MFCD00005114
CAS nummer: 2001-95-8
UN: 2811
ADR: 6.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78052be8082754804000cea112c9c3356d779702&vid=52adcfb29e375390b4270fa2a14206d676876171&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR