Supelco® PESTANAL® Organic Reference Standard, Clothianidin-d₃

Leverantör: Merck

56816-50MG
56816-50MGEA 2580 SEK
56816-50MG
Supelco® PESTANAL® Organic Reference Standard, Clothianidin-d₃
Standarder Organic Standards

  • GHS signal: GHS07, GHS09

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c621e0cbc3d39dbaed338a99543b6fc513f616c8&vid=c621e0cbc3d39dbaed338a99543b6fc513f616c8&prid=web&clid=SAPDAM

Order Now

SPECIFIKATIONER

Synonym(s) (E)-1-(2-Chloro-5-thiazolylmethyl)-3-(methyl-d₃)-2-nitroguanidine
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 1262776-24-8
MDL No. MFCD17019117
Molekylvikt 252,70
Faroklassbeskrivning H302 - H410
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR