VWR®, Snap Ring Vials ND11
Leverantör: VWR Collection
548-0012AEA 2475 SEK
548-0012A 548-1860A 548-1859A 548-0011A 548-0422A 548-1441A 548-1930A 548-0470A 548-0471A 548-0473A 548-0472A 548-8014A 548-8015A 548-8016A 548-2368A 548-2366A 548-2367A 548-1956A 548-1472A 548-0429A 548-0120A 548-0895A 548-0454A 548-0451A 548-1852A 548-2175A 548-2192A 548-2360A 548-2361A 548-3285A
VWR®, Snap Ring Vials ND11
Vialer
Snap ring ND11 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyethylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Wide opening
Snap ring ND11 vials constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear version with a flat bottom to ensure compatibility with inserts. Multiple cap colours mostly available with assembled septa are engineered to prevent sampleMore Product Information