Supelco® Ascentis® Express Phenyl-Hexyl HPLC and UHPLC Columns
Leverantör: Merck
53332-U
53334-U
53335-U
53336-U
53338-U
53341-U
53342-U
53343-U
53345-U
53346-U
53347-U
53348-U
53351-U
53352-U
53353-U
53524-U
53526-U
53531-U
53798-U
50442-U
50443-U
50446-U
50451-U
50454-U
50455-U
50456-U
50459-U
50464-U
50466-U
50469-U
50470-U
50472-U
50474-U
50477-U
50479-U
50482-U
50483-U
50487-U
50498-U
51600-U
51601-U
51603-U
51605-U
51608-U
51609-U
51611-U
51614-U
51616-U
51617-U
51618-U
51619-U
51623-U
53332-UEA
5340
SEK
53332-U
53334-U
53335-U
53336-U
53338-U
53341-U
53342-U
53343-U
53345-U
53346-U
53347-U
53348-U
53351-U
53352-U
53353-U
53524-U
53526-U
53531-U
53798-U
50442-U
50443-U
50446-U
50451-U
50454-U
50455-U
50456-U
50459-U
50464-U
50466-U
50469-U
50470-U
50472-U
50474-U
50477-U
50479-U
50482-U
50483-U
50487-U
50498-U
51600-U
51601-U
51603-U
51605-U
51608-U
51609-U
51611-U
51614-U
51616-U
51617-U
51618-U
51619-U
51623-U
Supelco® Ascentis® Express Phenyl-Hexyl HPLC and UHPLC Columns
Kromatografikolonn
SPECIFIKATIONER
Benämning | 2,7 µm columns | 5,0 µm columns | 2,0 µm columns |
Environmentally Preferable | |||
Applikation | Reversed-phase HPLC separations of peptides and polypeptides | Reversed-phase HPLC separations of peptides and polypeptides | Reversed-phase HPLC separations of peptides and polypeptides |
Kolbelastning | 7,1% | 5,2% | 6,3% |
Endcapped | Ja | Ja | Ja |
Max. tryck | 600 bar | 600 bar | 1000 bar |
pH område | 2 - 9 | 2 - 9 | 2 - 9 |
Surface area | 135 m²/g | 90 m²/g | 120 m²/g |
USP code | L11 | L11 | L11 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...