Supelco® Ascentis® Express ES-Cyano HPLC and UHPLC Columns
Leverantör: Merck
53468-U 53470-U 53472-U 53473-U 53475-U 53476-U 53478-U 53479-U 53481-U 53483-U 53484-U 53486-U 53489-U 53491-U 53492-U 53494-U 53495-U 53496-U 53497-U 50557-U 50558-U 50559-U 50562-U 50563-U 50564-U 50566-U 50567-U 50568-U 50569-U 50570-U 50574-U 50575-U 50577-U 50581-U 50583-U 50585-U 50588-U 50591-U 50592-U 50593-U 50597-U 51709-U 51712-U 51717-U 51724-U 51725-U 51727-U 51728-U 51729-U 51732-U 51734-U 51736-U 51739-U
53468-UEA 5340 SEK
53468-U 53470-U 53472-U 53473-U 53475-U 53476-U 53478-U 53479-U 53481-U 53483-U 53484-U 53486-U 53489-U 53491-U 53492-U 53494-U 53495-U 53496-U 53497-U 50557-U 50558-U 50559-U 50562-U 50563-U 50564-U 50566-U 50567-U 50568-U 50569-U 50570-U 50574-U 50575-U 50577-U 50581-U 50583-U 50585-U 50588-U 50591-U 50592-U 50593-U 50597-U 51709-U 51712-U 51717-U 51724-U 51725-U 51727-U 51728-U 51729-U 51732-U 51734-U 51736-U 51739-U
Supelco® Ascentis® Express ES-Cyano HPLC and UHPLC Columns
Kromatografikolonn
Ascentis® Express ES-Cyano high-speed, high-performance liquid chromatography columns are based on the highly efficient Fused-Core® particle design.
The Fused-core particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,5 µm thick porous shell.
The sterically protected, extensively endcapped diisopropyl-cyanopropylsilane stationary phase of Ascentis Express ES-Cyano provides a stable, reversed-phase packing that can be used for basic, acidic orMore Product Information