Saltsyra 36%, SLSI Selectipur® Electronic_industry

Leverantör: BASF Electronic Materials
Fara

Synonyms: Hydrochloric acid solution, Muriatic acid

52976033
52976033.EA 0 SEK
52976033.
Saltsyra 36%, SLSI Selectipur® Electronic_industry
Saltsyra

Product from BASF


Leveransinformation: This product is not available in all countries. Please check with your local VWR office or supplier.


Observera: This product is not available in all countries. Please check with your local VWR office or supplier.

Formel: HCl
MW: 36.46 g/mol
Smältpunkt: –30 °C
Densitet: 1.18 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
REACH: 01-2119484862-27

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR