Phenylarsine oxide, Calbiochem®

Leverantör: Merck Millipore Calbiochem

Synonyms: PAO

521000-250
521000-250EA 1150 SEK
521000-250
Phenylarsine oxide, Calbiochem®
Phenylarsine oxide
Formel: C₆H₅AsO
MW: 168.02 g/mol
MDL Number: MFCD00001990
CAS nummer: 637-03-6
EINECS: 211-275-3
UN: 3465
ADR: 6.1 III

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR