Biosafe benchtop ultracentrifuge System, Optima™ Max-XP
Leverantör: Beckman Coulter
A47882
521-4174EA 0 SEK
521-4174
Biosafe benchtop ultracentrifuge System, Optima™ Max-XP
Centrifuger Bordscentrifuger
The Optima MAX-XP's convenient touch-screen control features easy-to-read displays for gauging real-time speed, time, and temperature. The customisable user interface allows personalisation of easy-to-identify icons for up to twelve individuals, as well as sound and volume options. Multiple language capability: Access to an array of languages increases the Optima MAX-XP's functionality, anywhere in the world. From French and Spanish to Japanese, simplified Chinese and more, the Optima MAX-XP's ability to work in multiple languages enhances its value for laboratories around the world.