Bioprocessing centrifuges, refrigerated, floor standing, Thermo Scientific™ Sorvall™ BIOS 16 and Sorvall™ BIOS A
Leverantör: SORVALL
75007685 75007697 75007698 75007699
521-2685EA 0 SEK
521-2685 521-2686 521-2687 521-2688
Bioprocessing centrifuges, refrigerated, floor standing, Thermo Scientific™ Sorvall™ BIOS 16 and Sorvall™ BIOS A
Centrifuger Golvcentrifuger
These large capacity centrifuges are designed for high throughput bioprocessing and support a broad array of applications: Biologics, vaccines, biofuels, veterinary medicines, recombinant proteins and raw materials.
  • Glove-friendly interface with bright and highly visible display simplifies run set-up
  • Automatic 'front to back' centrifuge door opening and closing with the auto-door function
  • Auto-ID instant rotor identification: Immediate identification of a rotor when secured in the centrifuge chamber with rotor specifications automatically loaded into the
More Product Information