VWR® Micro Star 30R Microcentrifuge, High Speed, Universal, Refrigerated
Leverantör: VWR Collection
521-2574EA 0 SEK
521-2574 521-2575
VWR® Micro Star 30R Microcentrifuge, High Speed, Universal, Refrigerated
Centrifuger Mikrocentrifuger
The compact Micro Star 30R refrigerated centrifuge is an economic, versatile and easy to use cross-over centrifuge which combines the features of a high speed microcentrifuge and a multipurpose centrifuge. This quiet and smooth running centrifuge accommodates rotors for microtubes (24×1,5/2,0 ml), microplates and ¹⁵/₅₀ ml conical tubes as well as a 16-place rotor for 5 ml tubes and a swing-bucket rotor for six blood collection tubes. Ideal for use in life science, biotechnology laboratories and clinical laboratories.
  • Temperature range from –9 to +40 °C with pre-cooling function
  • Max.
More Product Information