Centrifuge, refrigerated, floor-standing, Cryofuge® 5500i
Leverantör: Thermo Scientific
75004471 75004478
521-1026EA 0 SEK
521-1026 HERA75004478
Centrifuge, refrigerated, floor-standing, Cryofuge® 5500i
Centrifuger Golvcentrifuger
Developed to meet the needs of users in a wide range of environments - including hospital blood banks and laboratories, satellite blood processing centers, and clinical reference labs - the versatile refrigerated centrifuge Cryofuge® 5500i provides exceptional processing power for a host of applications.
  • Optimal capacity for small to medium size blood processing centers
  • Can spin up to eight blood bags, in a wide range of bag configurations – including 500 ml quints with in-line filters
  • Effortless access with Soft-touch lid and easy-open/close mechanism
More Product Information