Phytochemical standard, Silychristin, analytical standard, Supelco®
Leverantör: Merck
51681-10MG
51681-10MGEA 0 SEK
51681-10MG
Phytochemical standard, Silychristin, analytical standard, Supelco®
Standarder Organic Standards
Silychristin is one of the congeners of flavonolignan silybin. It is usually derived from silymarin, which is extracted from the fruits of Silybum marianum (L.) Gaertn, and Asteraceae (Carduus marianus L. milk thistle).
Förpackning: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a21d78c752ed46b691afe4c6eedc286053cc9ef2&vid=a21d78c752ed46b691afe4c6eedc286053cc9ef2&prid=web&clid=SAPDAM