n-Hentriacontane, Supelco®

Leverantör: Merck


MQ100

51529-1G 51529-250MG
51529-1GEA 2170 SEK
51529-1G 51529-250MG
n-Hentriacontane, Supelco®
n-Hentriacontane
Formel: H₃C(CH₂)₂₉CH₃
MW: 436.85 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00048745
CAS nummer: 630-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1611d34e11fa4388bdb5de1874fb6d45b2a78123&vid=5638ee6769a590f94ae78e46d44c266c004477ac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR