VWR®, Syringe Filters
Leverantör: VWR Collection
514-1251EA 1200 SEK
514-1251 514-1249 514-1250 514-1240 514-1241 514-1243 514-1244 514-1245 514-1246 514-1247 514-1248 514-1252 514-1253 514-1254 514-1255 514-1256 514-1257 514-1258 514-1259 514-1260 514-1261 514-1262 514-1263 514-1264 514-1265 514-1266 514-1267 514-1268 514-1277 514-1269 514-1270 514-1271 514-1272 514-1273 514-1274 514-1275 514-1276 514-1278 514-1279 514-1280 514-1281
VWR®, Syringe Filters
Filter Sprutfilter
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
  • Sterile blister packed filters are ideal for sterile filtration
  • Non sterile syringe filters are excellent for HPLC applications
  • Biosafe is according to Class VI plastics tests
  • Available with nylon, PES, PTFE, PVDF, CA, RC membrane, and GF (Glass Fibre)
  • Available with hydrophilic or hydrophobic PTFE
  • Each filter has a female Luer lock inlet and a male slip Luer outlet
Nylon and PTFE membranes are ideal for sample preparation and small volume chemicalMore Product Information