Bottle filling bells, Nalgene™
Leverantör: Thermo Fisher Scientific
DS0390-0070
513-3901EA 780 SEK
513-3901
Bottle filling bells, Nalgene™
Flaskor Cellodlingsflaskor
Transparent, autoclavable filling bell, made of PC with PP tubing adapter, is for use in transferring sterile media aseptically from large containers to media bottles or culture vessels in microbiology and cell culture labs.
  • Autoclavable polypropylene tubing adapter accepts 6,3, 9,5 and 12,7 mm Int. Ø tubing
  • Bell is durable, non-toxic and will not shatter
  • Costs much less than glass
Note: For research use only; not for in vitro diagnosis or parenterals.