Glasfilter, gasfördelningsrör
Leverantör: ROBU GLASFILTER
19220 19221 19222 19250 19251 19252 19223 19224 19253 19254 19340 19341 19342 19343 19344
511-0700EA 780 SEK
511-0700 511-0701 511-0702 511-0710 511-0711 511-0712 511-0349 511-0350 511-0351 511-0352 511-0353 511-0354 511-0355 511-0356 511-0357
Glasfilter, gasfördelningsrör
Gasrör
These versatile and recyclable tubes offer excellent properties for diverse applications are made of borosilicate glass 3.3.
  • High resistance to most chemicals, such as solvents, acids and alkalis
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes