Analytical sieve shaker AS 200 jet
Leverantör: Retsch GmbH
30.027.0001
510-1379EA 0 SEK
510-1379
Analytical sieve shaker AS 200 jet
Skakapparater Skakapparater för siktar
The Retsch analytical air jet sieve shaker is particularly suitable for dispersion and deagglomeration of fine powders. Typical applications include construction materials, flour, spices, plastics and pharmaceuticals. The evaluation software EasySieve® can be used for automated evaluation and documentation.
  • Reproducible results due to Open Mesh Function which reduces the number of near-mesh particles
  • Quick start mode for easy operation
  • Short sieving times of 2 to 3 minutes
  • Silent operation due to integrated silencer
According to DIN EN ISO 9000 ff.