Kaliumdiväteortofosfat, anhydrous 99,5 Acidimetri, LiChropur™ HPLC, Supelco®

Leverantör: Merck

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

MQ100

5.43841.0100 5.43841.0250
5.43841.0100EA 1790 SEK
5.43841.0100 5.43841.0250
Kaliumdiväteortofosfat, anhydrous 99,5 Acidimetri, LiChropur™ HPLC, Supelco®
Kaliumdiväteortofosfat
Formel: KH₂PO₄
MW: 136.09 g/mol
Kokpunkt: 100
Smältpunkt: 253 °C
Densitet: 2.33 g/cm³ (21.5 °C)
MDL Number: MFCD00011401
CAS nummer: 7778-77-0
EINECS: 231-913-4
Merck Index: 13,07744

Order Now

Specifikation testresultat

Assay (acidometric) ≥99,5%
Identity Passes test
Ca (calcium) ≤0,50 ppm
Cu (copper) ≤0,005 ppm
Fe (iron) ≤0,10 ppm
Mg (magnesium) ≤0,50 ppm
Na (sodium) ≤20,0 ppm
HPLC gradient suitability test, chromophoric impurities based on 4'-hydroxyacetophenone ≤10 ppm
Chromophore purity ≥99,999%

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR