(2,4-Diklorfenoxi)ättiksyra

Leverantör: Merck
Fara

Synonyms: 2,4-D

49083
49083.EA 726 SEK
49083.
(2,4-Diklorfenoxi)ättiksyra
(2,4-Diklorfenoxi)ättiksyra
Formel: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04 g/mol
Kokpunkt: 346
Smältpunkt: 137…139 °C
Densitet: 1.563 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004300
CAS nummer: 94-75-7
EINECS: 202-361-1
Merck Index: 13,02825
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd852ae2ce44027c3e885b3f063c10354823a85f&vid=dd852ae2ce44027c3e885b3f063c10354823a85f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR