PCR-Cooler and PCR Racks
Leverantör: EPPENDORF
3881000.015 3881000.023 3881000.031 0030124.545
479-1160EA 1450 SEK
479-1160 479-1161 479-1162 713-0004
PCR-Cooler and PCR Racks
Kylare Kylare, bänkmodell
Eppendorf PCR-Racks for safe PCR sample storage/transport with color coding for easy identification - ideal for 0,1, 0,2, 0,5 ml tubes/strips. The PCR-Cooler keeps samples cool for 1hr+ post 2hrs in –20 °C, with temperature indicator for sample safety.
  • Handling system for sample set-up, protection, transport, and storage of sensitive samples - keeps your samples safe
  • Clear temperature indicator: Color of PCR-Cooler changes when temperature exceeds 7 °C
  • Accommodates PCR vessels as tubes, strips, or plates for flexible vessel usage
  • Dry incubation technology
More Product Information
Order Now
SPECIFIKATIONER
Environmentally Preferable Low Manufacturing Impact
Miljöcertificering Renewable Energy Certificate, ISO 14001