Cryogenic vials, CryoELITE®, WHEATON®
Leverantör: DWK Life Sciences
W985869 W985868 W985867 W985866 W985865 W985864 W985863 W985862 W985860 W985872 W985861 W985914 W985924 W985923 W985922 W985921 W985920 W985919 W985918 W985917 W985870 W985915 W985871 W985913 W985912 W985911 W985910 W985925 W985916 W985874
479-0975EA 6950 SEK
479-0975 479-0874 479-0873 479-0872 479-0871 479-0870 479-0868 479-0974 479-1135 479-0869 479-1136 479-1145 479-0980 479-0979 479-0899 479-0905 479-0904 479-0903 479-0902 479-0901 479-0976 479-0978 479-0977 479-1144 479-1143 479-1140 479-1139 479-0981 479-0900 479-2073
Cryogenic vials, CryoELITE®, WHEATON®
Vialer
CryoELITE® cryogenic vials are made from low binding, cryogenic grade virgin PP. They offer an unrivaled cap seal for the shipping and storage of precious biosamples. The patented cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handling.
  • Loctagon™ Vial Skirt provides stability in freestanding position
  • Screw cap can be easily removed with one hand
  • Coloured caps allow for colour coding
  • Round bottom vials can be centrifuged up to 17 000×g
Optional 2D data matrix barMore Product Information