Natriumhydroxid concentrate for dilution to 1.0 N i vattenlösning, StandARd® volumetrisk lösning, Macron Fine Chemicals™

Leverantör: Avantor Performance Materials
Fara

Synonyms: Natronlut, Kaustik soda

4693-09EA 4000 SEK
4693-09
Natriumhydroxid concentrate for dilution to 1.0 N i vattenlösning, StandARd® volumetrisk lösning, Macron Fine Chemicals™
Natriumhydroxid
Formel: NaOH
MW: 40 g/mol
Kokpunkt: 1390
Smältpunkt: 323
Densitet: 1.04 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now

Specifikation testresultat

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Normality 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR