Exsickator, underdel, DURAN®
Leverantör: DWK Life Sciences
247704604 247705703 247706108 247706605 247706905 247714605 247715704 247716109 247716606 247716906 247725705 247726101 247726607 247726907
467-1110EA 1350 SEK
467-1110 467-1115 467-1120 467-1125 467-1130 467-1210 467-1215 467-1220 467-1225 467-1230 467-1315 467-1320 467-1325 467-1330
Exsickator, underdel, DURAN®
Exsickatorer Exsickatorunderdel
DURAN®, clear borosilicate glass 3.3. Ideal for drying and drying processes, storage under vacuum and protection from the effects of air and moisture. Applications include cultivation of organisms, material research and corrosion testing, cleaning equipment for analysis of trace elements, gas and liquid reactions and they can be used as a mini cleanroom for chip technology.
  • Ökad säkerhet tack vare DURAN® borosilikat kvalitetsglas
  • Samtliga modeller har planslipad fläns
  • Hög resistens mot slitage, temperaturförändringar och kemikalier
Certifikat: AllMore Product Information