VWR®, Termostatbad
Leverantör: VWR Collection
462-0219EA 52300 SEK
462-0219 462-0218 462-0217 462-0216 462-0215 462-0222 462-0220 462-0221
VWR®, Termostatbad
Termostater Termostatbad, öppna system Bad med Inbyggda Termostater
Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Both are easy to navigate with large, intuitive displays and multiple communication options including USB-A and B, RS232/485, Ethernet and external temperature probe. The reservoir drain can be accessed by removing the front panel. All models feature user-adjustable high temperature safety cut-off points as well as over-temperature protection. The DuraTop™ surface is cool to the touch even when operating at high temperatures.
  • Arbetstemperatur från RT +10 till +200° C med
More Product Information