Cooling thermostats, Proline Kryomats
Leverantör: LAUDA
L001652 L001653 L001654 L001655 L001656 L001657 L001658 L001659
461-1296EA 222900 SEK
461-1296 461-1298 461-1301 461-1303 461-1297 461-1299 461-1302 461-1304
Cooling thermostats, Proline Kryomats
Termostater Termostatbad, slutna system Cirkulationskylare
The Proline Kryomats units are floor-standing, compact cooling thermostats and are fitted with the Command control head as standard. This features a back-lit, graphic LCD display with actual values in 0,1/0,01 or 0,001 °C resolution, data input via either cursor keys or numerical soft keys. The display provides clear text messages and graphical representation of sequential temperature curves and is menu-driven in four languages (German, English, French and Spanish). With large bath opening and volumes of up to 40 litres, this range is particularly suitable for internal applications.