VWR® ARISTAR®, Terbium Standard Solutions for ICP-MS
Leverantör: VWR Chemicals
456932QEA 3325 SEK
456932Q 85633.180
VWR® ARISTAR®, Terbium Standard Solutions for ICP-MS
Standarder Cationic Standards Terbium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis
Certifikat: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities