Kvicksilver 10.000 mg/l i utspädd salpetersyra (av Hg) ARISTAR® standard för ICP
Leverantör: VWR Chemicals
455672LEA 3375 SEK
455672L 455674N
Kvicksilver 10.000 mg/l i utspädd salpetersyra (av Hg) ARISTAR® standard för ICP
Standarder Cationic Standards Mercury Standards
Hg in HNO₃ 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.